3-Carbamothioylpiperidine, N1-BOC protected
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl 3-carbamothioylpiperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-carbamothioylpiperidine, 1-(tert-Butoxycarbonyl)piperidine-3-thiocarboxamide
OR59984-1G
OR59984-5G
APOSOR59984-1GEA
6250
CZK
APOSOR59984-1G
APOSOR59984-5G
3-Carbamothioylpiperidine, N1-BOC protected
3-Carbamothioylpiperidine, N1-BOC protected
Vzorec:
C₁₁H₂₀N₂O₂S Bod tání: 134…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD10700060 CAS číslo: 274682-80-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...