Tisk…

5-Phenylpyridine-2-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Phenylpicolinic acid

OR59922-1G OR59922-250MG
APOSOR59922-1GEA 8700 CZK
APOSOR59922-1G APOSOR59922-250MG
5-Phenylpyridine-2-carboxylic acid 95%
5-Phenylpyridine-2-carboxylic acid
Vzorec: C₁₂H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD04114189
CAS číslo: 75754-04-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR