3,4-Dihydro-6-nitro-1H-quinolin-2-one
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Nitro-3,4-dihydroquinolin-2(1H)-one
OR59827-25G
OR59827-5G
APOSOR59827-25GEA
6290
CZK
APOSOR59827-25G
APOSOR59827-5G
3,4-Dihydro-6-nitro-1H-quinolin-2-one
3,4-Dihydro-6-nitro-1H-quinolin-2-one
Vzorec:
C₉H₈N₂O₃ MW: 192,171 g/mol Bod tání: 201…202 °C Storage Temperature: Freezer |
MDL Number:
MFCD00559317 CAS číslo: 22246-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...