Tisk…

2,5-Dimethylfuran-3-sulphonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR59789-1G OR59789-250MG
APOSOR59789-1GEA 10290 CZK
APOSOR59789-1G APOSOR59789-250MG
2,5-Dimethylfuran-3-sulphonyl chloride 97%
2,5-Dimethylfuran-3-sulphonyl chloride
Vzorec: C₆H₇ClO₃S
Bod varu: 244…245 °C
Storage Temperature: Ambient
MDL Number: MFCD04115370
CAS číslo: 166964-26-7
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR