Tisk…

4-{[Methoxy(methyl)carbamoyl]amino}benzeneboronic acid, pinacol ester 98%

Dodavatel: Apollo Scientific

Synonyms: 4-(3-Methoxy-3-methylureido)benzeneboronic acid, pinacol ester

OR5973-1G OR5973-250MG
APOSOR5973-1GEA 10780 CZK
APOSOR5973-1G APOSOR5973-250MG
4-{[Methoxy(methyl)carbamoyl]amino}benzeneboronic acid, pinacol ester 98%
4-{[Methoxy(methyl)carbamoyl]amino}benzeneboronic acid, pinacol ester
Vzorec: C₁₅H₂₃BN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD09027291
CAS číslo: 874297-84-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR