Tisk…

1-[4-(Chloromethyl)phenyl]-1H-pyrazole 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR59436-1G OR59436-5G
APOSOR59436-1GEA 1950 CZK
APOSOR59436-1G APOSOR59436-5G
1-[4-(Chloromethyl)phenyl]-1H-pyrazole 95%
1-[4-(Chloromethyl)phenyl]-1H-pyrazole
Vzorec: C₁₀H₉ClN₂
Storage Temperature: Ambient
MDL Number: MFCD07366379
CAS číslo: 143426-52-2
UN: 1759
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR