Methyl 6-(pyrrolidin-1-yl)nicotinate 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR59370-1G
OR59370-250MG
OR59370-5G
APOSOR59370-1GEA
1870
CZK
APOSOR59370-1G
APOSOR59370-250MG
APOSOR59370-5G
Methyl 6-(pyrrolidin-1-yl)nicotinate 97%
Methyl 6-(pyrrolidin-1-yl)nicotinate
Vzorec:
C₁₁H₁₄N₂O₂ Bod tání: 119…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664433 CAS číslo: 210963-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...