Tisk…

2-Chloro-4-(4-methoxyphenyl)pyrimidine

Dodavatel: Apollo Scientific
Varování

OR59362-1G OR59362-500MG
APOSOR59362-1GEA 9290 CZK
APOSOR59362-1G APOSOR59362-500MG
2-Chloro-4-(4-methoxyphenyl)pyrimidine
2-Chloro-4-(4-methoxyphenyl)pyrimidine
Vzorec: C₁₁H₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD09861469
CAS číslo: 75634-04-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR