Tisk…

4-[(5-Bromopyridin-2-yl)carbonyl]morpholine

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-2-(morpholin-4-ylcarbonyl)pyridine, (5-Bromopyridin-2-yl)(morpholino)methanone, (5-Bromopyridine-2-yl )(morpholino)methanone, (5-Bromopyridin-2-yl)(morpholin-4-yl)methanone

OR59349-1G OR59349-250MG
APOSOR59349-1GEA 5200 CZK
APOSOR59349-1G APOSOR59349-250MG
4-[(5-Bromopyridin-2-yl)carbonyl]morpholine
4-[(5-Bromopyridin-2-yl)carbonyl]morpholine
Vzorec: C₁₀H₁₁BrN₂O₂
MW: 271,111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878428
CAS číslo: 957063-06-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR