Tisk…

Ethyl 2-sulphamoylbenzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(Ethoxycarbonyl)benzenesulphonamide

OR5787-100G OR5787-25G OR5787-5G
APOSOR5787-100GEA 2690 CZK
APOSOR5787-100G APOSOR5787-25G APOSOR5787-5G
Ethyl 2-sulphamoylbenzoate
Ethyl 2-sulphamoylbenzoate
Vzorec: C₉H₁₁NO₄S
MW: 229.253 g/mol
Bod tání: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00455715
CAS číslo: 59777-72-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR