3-Chloro-4-(hydrazinocarbonyl)benzeneboronic acid 97%
Dodavatel: Apollo Scientific
OR5760-1G
OR5760-5G
APOSOR5760-1GEA
6720
CZK
APOSOR5760-1G
APOSOR5760-5G
3-Chloro-4-(hydrazinocarbonyl)benzeneboronic acid 97%
3-Chloro-4-(hydrazinocarbonyl)benzeneboronic acid
Vzorec:
C₇H₈BClN₂O₃ MW: 214.414 g/mol Bod tání: 144…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD07363764 CAS číslo: 850589-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...