4-Bromo-2-nitrophenylhydrazine 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR5743-1G
OR5743-5G
OR5743-250MG
APOSOR5743-1GEA
2940
CZK
APOSOR5743-1G
APOSOR5743-5G
APOSOR5743-250MG
4-Bromo-2-nitrophenylhydrazine 96%
4-Bromo-2-nitrophenylhydrazine
Vzorec:
C₆H₆BrN₃O₂ MW: 232,035 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03788274 CAS číslo: 59488-34-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...