Tisk…

1-Benzyl-4-hydrazinopiperidine dihydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Hydrazino-1-(phenylmethyl)piperidine dihydrochloride, (1-Benzylpiperidin-4-yl)hydrazine dihydrochloride

OR5649-1G OR5649-250MG
APOSOR5649-1GEA 11810 CZK
APOSOR5649-1G APOSOR5649-250MG
1-Benzyl-4-hydrazinopiperidine dihydrochloride
1-Benzyl-4-hydrazinopiperidine dihydrochloride
Vzorec: C₁₂H₁₉N₃·2ClH
MW: 278.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035362
CAS číslo: 83949-42-2
EINECS: 281-403-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR