Diethyl 1H-imidazole-4,5-dicarboxylate 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR5618-25G
OR5618-5G
APOSOR5618-25GEA
10320
CZK
APOSOR5618-25G
APOSOR5618-5G
Diethyl 1H-imidazole-4,5-dicarboxylate 97%
Diethyl 1H-imidazole-4,5-dicarboxylate
Vzorec:
C₉H₁₂N₂O₄ Bod tání: 149…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00723829 CAS číslo: 1080-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...