Tisk…

(7-Bromobenzofuran-2-yl)methanol

Dodavatel: Apollo Scientific
Varování

OR55740-1G OR55740-250MG
APOSOR55740-1GEA 15480 CZK
APOSOR55740-1G APOSOR55740-250MG
(7-Bromobenzofuran-2-yl)methanol
(7-Bromobenzofuran-2-yl)methanol
Vzorec: C₉H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD21233818
CAS číslo: 1467056-12-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR