2-(Cyclopropylmethyl)-1,3-thiazole-5-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR55735-100MG
OR55735-1G
OR55735-250MG
APOSOR55735-100MGEA
19010
CZK
APOSOR55735-100MG
APOSOR55735-1G
APOSOR55735-250MG
2-(Cyclopropylmethyl)-1,3-thiazole-5-carboxylic acid 95%
2-(Cyclopropylmethyl)-1,3-thiazole-5-carboxylic acid
Vzorec:
C₈H₉NO₂S MW: 183,228 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23912286 CAS číslo: 1526717-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...