Tisk…

3-Bromo-2,6-dichlorophenol 97%

Dodavatel: Apollo Scientific
Varování

OR55726-1G
APOSOR55726-1GEA 11870 CZK
APOSOR55726-1G
3-Bromo-2,6-dichlorophenol 97%
3-Bromo-2,6-dichlorophenol
Vzorec: C₆H₃BrCl₂O
MW: 241,897 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17011993
CAS číslo: 116668-74-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR