Tisk…

2-Amino-7-naphthol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-7-hydroxynaphthalene

OR55723-100MG OR55723-250MG OR55723-500MG
APOSOR55723-100MGEA 7250 CZK
APOSOR55723-100MG APOSOR55723-250MG APOSOR55723-500MG
2-Amino-7-naphthol 95%
2-Amino-7-naphthol
Vzorec: C₁₀H₉NO
Storage Temperature: Ambient
MDL Number: MFCD06252604
CAS číslo: 93-36-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR