Ethyl 4-bromo-1-benzofuran-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR55704-1G
OR55704-250MG
APOSOR55704-1GEA
11380
CZK
APOSOR55704-1G
APOSOR55704-250MG
Ethyl 4-bromo-1-benzofuran-2-carboxylate
Ethyl 4-bromo-1-benzofuran-2-carboxylate
Vzorec:
C₁₁H₉BrO₃ MW: 269,091 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16038163 CAS číslo: 177735-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...