1-Phenyl-1,2,4-triazole-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR55688-1G
OR55688-250MG
APOSOR55688-1GEA
8900
CZK
APOSOR55688-1G
APOSOR55688-250MG
1-Phenyl-1,2,4-triazole-3-carboxylic acid
1-Phenyl-1,2,4-triazole-3-carboxylic acid
Vzorec:
C₉H₇N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09260327 CAS číslo: 24036-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...