6-Bromobenzofuran-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR55679-1G
OR55679-250MG
APOSOR55679-1GEA
5460
CZK
APOSOR55679-1G
APOSOR55679-250MG
6-Bromobenzofuran-2-carboxylic acid
6-Bromobenzofuran-2-carboxylic acid
Vzorec:
C₉H₅BrO₃ Storage Temperature: Ambient |
MDL Number:
MFCD03012913 CAS číslo: 439107-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...