2,6-Dibromo-N-methylbenzene-1-sulphonamide
Dodavatel: Apollo Scientific
|
Varování
|
OR55678-1G
OR55678-250MG
APOSOR55678-1GEA
4770
CZK
APOSOR55678-1G
APOSOR55678-250MG
2,6-Dibromo-N-methylbenzene-1-sulphonamide
2,6-Dibromo-N-methylbenzene-1-sulphonamide
Vzorec:
C₇H₇Br₂NO₂S MW: 329,009 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30329437 CAS číslo: 1864153-96-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...