Tisk…

2-Bromo-1-(2-cyclopropylphenyl)ethanone

Dodavatel: Apollo Scientific
Nebezpečí

OR55674-1G OR55674-250MG
APOSOR55674-1GEA 3110 CZK
APOSOR55674-1G APOSOR55674-250MG
2-Bromo-1-(2-cyclopropylphenyl)ethanone
2-Bromo-1-(2-cyclopropylphenyl)ethanone
Vzorec: C₁₁H₁₁BrO
MW: 239.108 g/mol
Storage Temperature: Freezer
MDL Number: MFCD29040961
CAS číslo: 1936493-49-0
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR