(5-Methyl-1,3-oxazol-4-yl)methanol
Dodavatel: Apollo Scientific
|
Varování
|
OR55671-1G
OR55671-250MG
APOSOR55671-1GEA
12140
CZK
APOSOR55671-1G
APOSOR55671-250MG
(5-Methyl-1,3-oxazol-4-yl)methanol
(5-Methyl-1,3-oxazol-4-yl)methanol
Vzorec:
C₅H₇NO₂ MW: 113,115 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16556111 CAS číslo: 874821-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...