4-Bromo-2-cyclopropylphenol
Dodavatel: Apollo Scientific
|
Varování
|
OR55665-1G
OR55665-250MG
APOSOR55665-1GEA
15710
CZK
APOSOR55665-1G
APOSOR55665-250MG
4-Bromo-2-cyclopropylphenol
4-Bromo-2-cyclopropylphenol
Vzorec:
C₉H₉BrO MW: 213,071 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD24715363 CAS číslo: 1043871-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...