Tisk…

4-Iodobenzo[D]isoxazol-3-amine

Dodavatel: Apollo Scientific
Nebezpečí

OR55634-1G OR55634-250MG
APOSOR55634-1GEA 3310 CZK
APOSOR55634-1G APOSOR55634-250MG
4-Iodobenzo[D]isoxazol-3-amine
4-Iodobenzo[D]isoxazol-3-amine
Vzorec: C₇H₅IN₂O
MW: 260,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040329
CAS číslo: 1012367-55-3
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR