Tisk…

5-Benzyloxy-1H-indole-2-boronic acid, N-BOC protected 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(Benzyloxy)-1-(tert-butoxycarbonyl)-1H-indole-2-boronic acid, (5-(Benzyloxy)-1-(tert-butoxycarbonyl)-1H-indol-2-yl)boronic acid

OR5563-1G OR5563-5G
APOSOR5563-1GEA 1740 CZK
APOSOR5563-1G APOSOR5563-5G
5-Benzyloxy-1H-indole-2-boronic acid, N-BOC protected 98%
5-Benzyloxy-1H-indole-2-boronic acid, N-BOC protected
Vzorec: C₂₀H₂₂BNO₅
MW: 367,203 g/mol
Bod tání: 106…110 °C
Storage Temperature: Freezer
MDL Number: MFCD06659824
CAS číslo: 850568-62-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR