Tisk…

3-(Oxan-4-yl)azetidine hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

OR55621-100MG OR55621-250MG
APOSOR55621-100MGEA 36040 CZK
APOSOR55621-100MG APOSOR55621-250MG
3-(Oxan-4-yl)azetidine hydrochloride 95%
3-(Oxan-4-yl)azetidine hydrochloride
Vzorec: C₈H₁₅NO·ClH
MW: 177,672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20483682
CAS číslo: 251569-98-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR