Tisk…

2-Benzyloxy-5-chlorophenyl acetic acid 95%

Dodavatel: Apollo Scientific
Varování

OR55609-1G OR55609-250MG
APOSOR55609-1GEA 13230 CZK
APOSOR55609-1G APOSOR55609-250MG
2-Benzyloxy-5-chlorophenyl acetic acid 95%
2-Benzyloxy-5-chlorophenyl acetic acid
Vzorec: C₁₅H₁₃ClO₃
MW: 276,715 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664747
CAS číslo: 149428-98-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR