Tisk…

6-Bromo-2-isopropyl-2H-indazole 95%

Dodavatel: Apollo Scientific
Varování

OR55602-1G OR55602-250MG
APOSOR55602-1GEA 24570 CZK
APOSOR55602-1G APOSOR55602-250MG
6-Bromo-2-isopropyl-2H-indazole 95%
6-Bromo-2-isopropyl-2H-indazole
Vzorec: C₁₀H₁₁BrN₂
MW: 239,112 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31692108
CAS číslo: 1683546-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR