4-Bromo-2-iodobenzoic acid 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR55547-100G
OR55547-25G
OR55547-5G
APOSOR55547-100GEA
9590
CZK
APOSOR55547-100G
APOSOR55547-25G
APOSOR55547-5G
4-Bromo-2-iodobenzoic acid 95+%
4-Bromo-2-iodobenzoic acid
Vzorec:
C₇H₄BrIO₂ MW: 326,914 g/mol Bod tání: 183…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD12767500 CAS číslo: 1133123-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...