Tisk…

2-Chloro-4-methoxybenzylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (2-Chloro-4-methoxyphenyl)methanamine

OR55545-1G OR55545-5G
APOSOR55545-1GEA 5790 CZK
APOSOR55545-1G APOSOR55545-5G
2-Chloro-4-methoxybenzylamine 97%
2-Chloro-4-methoxybenzylamine
Vzorec: C₈H₁₀ClNO
MW: 171,624 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06213373
CAS číslo: 247569-72-8
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR