2,4-Bis(methylsulphonyl)benzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR55491-1G
OR55491-5G
APOSOR55491-1GEA
7080
CZK
APOSOR55491-1G
APOSOR55491-5G
2,4-Bis(methylsulphonyl)benzaldehyde
2,4-Bis(methylsulphonyl)benzaldehyde
Vzorec:
C₉H₁₀O₅S₂ MW: 262.303 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28962763 CAS číslo: 1845690-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...