Tisk…

4-(Trichloromethyl)benzoyl chloride, Technical Grade

Dodavatel: Apollo Scientific
Nebezpečí

OR55473-1G OR55473-250MG OR55473-5G
APOSOR55473-1GEA 11540 CZK
APOSOR55473-1G APOSOR55473-250MG APOSOR55473-5G
4-(Trichloromethyl)benzoyl chloride, Technical Grade
4-(Trichloromethyl)benzoyl chloride
Vzorec: C₈H₄Cl₄O
MW: 257.929 g/mol
Bod tání: 27…29 °C
Storage Temperature: Ambient
MDL Number: MFCD25460286
CAS číslo: 14815-86-2
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR