6-Bromoisoquinoline-1-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
OR55470-1G
OR55470-250MG
APOSOR55470-1GEA
12240
CZK
APOSOR55470-1G
APOSOR55470-250MG
6-Bromoisoquinoline-1-carbonitrile
6-Bromoisoquinoline-1-carbonitrile
Vzorec:
C₁₀H₅BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD18633063 CAS číslo: 1082674-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...