Tisk…

(2R)-2-(Methylamino)propan-1-ol 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR55460-100MG OR55460-1G OR55460-250MG
APOSOR55460-100MGEA 4400 CZK
APOSOR55460-100MG APOSOR55460-1G APOSOR55460-250MG
(2R)-2-(Methylamino)propan-1-ol 95%
(2R)-2-(Methylamino)propan-1-ol
Vzorec: C₄H₁₁NO
MW: 89,136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19218361
CAS číslo: 184593-05-3
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR