5-Bromo-7-iodobenzofuran
Dodavatel: Apollo Scientific
|
Varování
|
OR55454-250MG
OR55454-1G
APOSOR55454-250MGEA
6620
CZK
APOSOR55454-250MG
APOSOR55454-1G
5-Bromo-7-iodobenzofuran
5-Bromo-7-iodobenzofuran
Vzorec:
C₈H₄BrIO MW: 322.925 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31619571 CAS číslo: 2227272-90-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...