Tisk…

4-Bromo-5-iodo-2-nitroaniline 95%

Dodavatel: Apollo Scientific
Varování

OR55450-1G OR55450-250MG
APOSOR55450-1GEA 18620 CZK
APOSOR55450-1G APOSOR55450-250MG
4-Bromo-5-iodo-2-nitroaniline 95%
4-Bromo-5-iodo-2-nitroaniline
Vzorec: C₆H₄BrIN₂O₂
MW: 342,917 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31543536
CAS číslo: 2090498-07-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR