5-Chloro-2-hydroxyphenylacetic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(5-Chloro-2-hydroxyphenyl)acetic acid
OR55449-100MG
OR55449-250MG
OR55449-50MG
APOSOR55449-100MGEA
5760
CZK
APOSOR55449-100MG
APOSOR55449-250MG
APOSOR55449-50MG
5-Chloro-2-hydroxyphenylacetic acid 95%
5-Chloro-2-hydroxyphenylacetic acid
Vzorec:
C₈H₇ClO₃ MW: 186,592 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11840335 CAS číslo: 24161-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...