Tisk…

5-Chlorobenzofuran-2(3H)-one 97%

Dodavatel: Apollo Scientific

OR55448-250MG OR55448-1G
APOSOR55448-250MGEA 0 CZK
APOSOR55448-250MG APOSOR55448-1G
5-Chlorobenzofuran-2(3H)-one 97%
5-Chlorobenzofuran-2(3H)-one
Vzorec: C₈H₅ClO₂
MW: 168,577 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09842447
CAS číslo: 28033-47-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR