Tisk…

2-Chloro-5-hydrazinopyrazine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR55438-100MG OR55438-1G OR55438-500MG
APOSOR55438-100MGEA 3940 CZK
APOSOR55438-100MG APOSOR55438-1G APOSOR55438-500MG
2-Chloro-5-hydrazinopyrazine hydrochloride
2-Chloro-5-hydrazinopyrazine hydrochloride
Vzorec: C₄H₅ClN₄·ClH
MW: 181,023 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31619570
CAS číslo: 2227272-59-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR