Tisk…

8-Bromo-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1,-e]isothiazole 4-oxide 95%

Dodavatel: Apollo Scientific
Varování

OR55426-1G OR55426-250MG
APOSOR55426-1GEA 28730 CZK
APOSOR55426-1G APOSOR55426-250MG
8-Bromo-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1,-e]isothiazole 4-oxide 95%
8-Bromo-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1,-e]isothiazole 4-oxide
Vzorec: C₁₀H₁₀BrNOS
MW: 272,162 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31621090
CAS číslo: 2169310-95-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR