(3R,4R)-Pyrrolidine-3,4-diol 97%
Dodavatel: Apollo Scientific
OR55391-250MG
OR55391-1G
APOSOR55391-250MGEA
12440
CZK
APOSOR55391-250MG
APOSOR55391-1G
(3R,4R)-Pyrrolidine-3,4-diol 97%
(3R,4R)-Pyrrolidine-3,4-diol
Vzorec:
C₄H₉NO₂ MW: 103,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08686736 CAS číslo: 186393-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...