Tisk…

3-Bromo-2-chloro-6-iodobenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

OR55383-1G OR55383-250MG OR55383-5G
APOSOR55383-1GEA 19840 CZK
APOSOR55383-1G APOSOR55383-250MG APOSOR55383-5G
3-Bromo-2-chloro-6-iodobenzoic acid 95%
3-Bromo-2-chloro-6-iodobenzoic acid
Vzorec: C₇H₃BrClIO₂
MW: 361,359 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619567
CAS číslo: 2091247-87-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR