Tisk…

2,5-Dibromothiophene-3-carbonitrile

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,5-Dibromothiophene-3-carbonitrile

OR55333-250MG OR55333-1G
APOSOR55333-250MGEA 14520 CZK
APOSOR55333-250MG APOSOR55333-1G
2,5-Dibromothiophene-3-carbonitrile
2,5-Dibromothiophene-3-carbonitrile
Vzorec: C₅HBr₂NS
MW: 266,941 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22578679
CAS číslo: 18792-01-3
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR