Tisk…

5-Bromo-7-chloro-1,2,3,4-tetrahydroisoquinoline

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-7-chloro-1,2,3,4-tetrahydroisoquinoline

OR55276-250MG OR55276-1G OR55276-5G
APOSOR55276-250MGEA 3190 CZK
APOSOR55276-250MG APOSOR55276-1G APOSOR55276-5G
5-Bromo-7-chloro-1,2,3,4-tetrahydroisoquinoline
5-Bromo-7-chloro-1,2,3,4-tetrahydroisoquinoline
Vzorec: C₉H₉BrClN
MW: 246,531 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24129966
CAS číslo: 1522580-66-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR