N-Isopropylglycine, N-BOC protected
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N-Boc-N-Isopropylamino-acetic acid, 2-[(2-Methylpropan-2-yl)oxycarbonyl-propan-2-ylamino]acetic acid
OR55269-1G
OR55269-5G
APOSOR55269-1GEA
5130
CZK
APOSOR55269-1G
APOSOR55269-5G
N-Isopropylglycine, N-BOC protected
N-Isopropylglycine, N-BOC protected
Vzorec:
C₁₀H₁₉NO₄ MW: 217,262 g/mol Bod tání: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689645 CAS číslo: 154509-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...