3-Chloro-2-methyl-6-nitroaniline 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR55265-1G
OR55265-250MG
OR55265-5G
APOSOR55265-1GEA
1870
CZK
APOSOR55265-1G
APOSOR55265-250MG
APOSOR55265-5G
3-Chloro-2-methyl-6-nitroaniline 98%
3-Chloro-2-methyl-6-nitroaniline
Vzorec:
C₇H₇ClN₂O₂ MW: 186,596 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09026962 CAS číslo: 51123-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...