Tisk…

2'-Chloro-5'-iodoacetophenone

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(2-Chloro-5-iodophenyl)ethanone

OR55246-1G OR55246-250MG
APOSOR55246-1GEA 4960 CZK
APOSOR55246-1G APOSOR55246-250MG
2'-Chloro-5'-iodoacetophenone
2'-Chloro-5'-iodoacetophenone
Vzorec: C₈H₆ClIO
MW: 280,49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22493253
CAS číslo: 100479-73-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR