2-Methyl-1-propyl-1,3-benzodiazole-5-carboxylic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR55244-10G
OR55244-1G
OR55244-5G
APOSOR55244-10GEA
22390
CZK
APOSOR55244-10G
APOSOR55244-1G
APOSOR55244-5G
2-Methyl-1-propyl-1,3-benzodiazole-5-carboxylic acid 98%
2-Methyl-1-propyl-1,3-benzodiazole-5-carboxylic acid
Vzorec:
C₁₂H₁₄N₂O₂ MW: 218,252 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11103016 CAS číslo: 68740-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...